1-Methyl-5-nitro-1,2,3-triazole
Lieferant: Apollo Scientific
APOSOR470666-1GEA
601
EUR
APOSOR470666-1G
1-Methyl-5-nitro-1,2,3-triazole
1-Methyl-5-nitro-1,2,3-triazole
Formel:
C₃H₄N₄O₂ Molekulargewicht: 128,089 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD26045236 CAS-Nummer: 1204736-25-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...