4-Ethoxy-3-methylbenzoic acid
Lieferant: Apollo Scientific
APOSOR470796-5GEA
601
EUR
APOSOR470796-5G
4-Ethoxy-3-methylbenzoic acid
4-Ethoxy-3-methylbenzoic acid
Formel:
C₁₀H₁₂O₃ Molekulargewicht: 180,2 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD13920122 CAS-Nummer: 92315-60-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...