2-Amino-1,6-dimethylbenzimidazole

Lieferant: Apollo Scientific

APOSOR470825-1GEA 794 EUR
APOSOR470825-1G
2-Amino-1,6-dimethylbenzimidazole
2-Amino-1,6-dimethylbenzimidazole
Formel: C₉H₁₁N₃
Molekulargewicht: 161,204 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD20703217
CAS-Nummer: 945023-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR