4-[(Isopropylamino)sulphonyl]benzoic acid

Lieferant: Apollo Scientific

APOSOR470989-5GEA 713 EUR
APOSOR470989-5G
4-[(Isopropylamino)sulphonyl]benzoic acid
4-[(Isopropylamino)sulphonyl]benzoic acid
Formel: C₁₀H₁₃NO₄S
Molekulargewicht: 243,28 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06355943
CAS-Nummer: 10252-66-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR