Ethyl 2-(Benzofuran-5-yl)acetate
Lieferant: Apollo Scientific
APOSOR471141-1GEA
397
EUR
APOSOR471141-1G
Ethyl 2-(Benzofuran-5-yl)acetate
Ethyl 2-(Benzofuran-5-yl)acetate
Formel:
C₁₂H₁₂O₃ Molekulargewicht: 204,222 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD16036979 CAS-Nummer: 142935-51-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...