(E)-4-(Dimethylamino)-N-[4-(hydroxymethyl)phenyl]-2-butenamide hydrochloride

Lieferant: Apollo Scientific

APOSOR471514-250MGEA 713 EUR
APOSOR471514-250MG
(E)-4-(Dimethylamino)-N-[4-(hydroxymethyl)phenyl]-2-butenamide hydrochloride
(E)-4-(Dimethylamino)-N-[4-(hydroxymethyl)phenyl]-2-butenamide hydrochloride
Formel: C₁₃H₁₈N₂O₂·ClH
Molekulargewicht: 270,755 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD29049240
CAS-Nummer: 2006281-61-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR