5-DTAF hydrochloride

Lieferant: Apollo Scientific

APOSBID2096-100MGEA 154 EUR
APOSBID2096-100MG APOSBID2096-500MG
5-DTAF hydrochloride
5-DTAF hydrochloride

Fluorescein derivative with amino reactivity.

Formel: C₂₃H₁₂Cl₂N₄O₅·HCl
Molekulargewicht: 531,8 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00051407
CAS-Nummer: 21811-74-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR