L(+)-Lyxose
|
Achtung
|
APOSBIFS0110-10GEA
0
EUR
APOSBIFS0110-10G
APOSBIFS0110-5G
L(+)-Lyxose
L(+)-Lyxose
Formel:
C₅H₁₀O₅ Molekulargewicht: 150,13 g/mol Schmelzpunkt: 105…106 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00064111 CAS-Nummer: 1949-78-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...