Bleiantimonid 99+%

APOSIN2203-5GEA 29.2 EUR
APOSIN2203-5G APOSIN2203-25G
Bleiantimonid 99+%
Lead antimonide
Formel: PbSb
Molekulargewicht: 328,96 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00151190
CAS-Nummer: 12266-38-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR