Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')yttrium(III) 99,9%
APOSIN6498-10GEA
0
EUR
APOSIN6498-10G
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')yttrium(III) 99,9%
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')yttrium(III)
Formel:
C₃₃H₅₇O₆Y Molekulargewicht: 638,708 g/mol Schmelzpunkt: 170…175 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00015713 CAS-Nummer: 15632-39-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...