4-[(Trimethylsilyl)ethinyl]benzaldehyd
APOSOR01513-500MGEA
0
EUR
APOSOR01513-500MG
4-[(Trimethylsilyl)ethinyl]benzaldehyd
4-[(Trimethylsilyl)ethinyl]benzaldehyd
Formel:
C₁₂H₁₄OSi Molekulargewicht: 202,33 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02093765 CAS-Nummer: 77123-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...