2-Thiophen-2-yl-piperazine
APOSOR01614-50MGEA
0
EUR
APOSOR01614-50MG
2-Thiophen-2-yl-piperazine
2-Thiophen-2-yl-piperazine
Formel:
C₈H₁₂N₂S Molekulargewicht: 168,26 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06245415 CAS-Nummer: 85803-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...