5-(Chlormethyl)-3-(4-chlorphenyl)-1,2,4-oxadiazol 95+%
APOSOR0254-250MGEA
0
EUR
APOSOR0254-250MG
APOSOR0254-1G
5-(Chlormethyl)-3-(4-chlorphenyl)-1,2,4-oxadiazol 95+%
5-(Chloromethyl)-3-(4-chlorophenyl)-1,2,4-oxadiazole
Formel:
C₉H₆Cl₂N₂O Schmelzpunkt: 56 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00119077 CAS-Nummer: 57238-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...