2,3-Dichlorbenzamidoxim

APOSOR0339-1GEA 0 EUR
APOSOR0339-1G APOSOR0339-250MG
2,3-Dichlorbenzamidoxim
2,3-Dichlorbenzamidoxim
Formel: C₇H₆Cl₂N₂O
Schmelzpunkt: 141…144 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01567248
CAS-Nummer: 261761-55-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR