3-(Aminomethyl)phenylboronsäure Hydrochlorid 95%
|
Achtung
|
APOSOR10329-1GEA
0
EUR
APOSOR10329-1G
APOSOR10329-5G
APOSOR10329-25G
3-(Aminomethyl)phenylboronsäure Hydrochlorid 95%
3-(Aminomethyl)phenylboronsäure Hydrochlorid
Formel:
C₇H₁₁BClNO₂ Molekulargewicht: 187,43 g/mol Schmelzpunkt: 248…250 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD02093050 CAS-Nummer: 352525-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...