5-Amino-3-brom-1H-indazol
Lieferant: Apollo Scientific
|
Gefahr
|
APOSOR110096-1GEA
0
EUR
APOSOR110096-1G
5-Amino-3-brom-1H-indazol
5-Amino-3-bromo-1H-indazole
Formel:
C₇H₆BrN₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07781655 CAS-Nummer: 478837-59-1 UN-Nummer: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...