tert-Butylisocyanacetat
|
Achtung
|
APOSOR11142-1MLEA
0
EUR
APOSOR11142-1ML
APOSOR11142-5ML
tert-Butylisocyanacetat
tert-Butylisocyanacetat
Formel:
C₇H₁₁NO₂ Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD00042952 CAS-Nummer: 2769-72-4 UN-Nummer: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...