4-(4-Bromphenyl)-2-methylpyrimidin
APOSOR111500-1GEA
0
EUR
APOSOR111500-1G
4-(4-Bromphenyl)-2-methylpyrimidin
4-(4-Bromophenyl)-2-methylpyrimidine
Formel:
C₁₁H₉BrN₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02089754 CAS-Nummer: 499785-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...