Methyl-L-prolinathydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR11387-25GEA
0
EUR
APOSOR11387-25G
APOSOR11387-100G
Methyl-L-prolinathydrochlorid
L-Proline methyl ester hydrochloride
Formel:
C₆H₁₁NO₂·HCl Molekulargewicht: 165,6 g/mol Schmelzpunkt: 69…78 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00012708 CAS-Nummer: 2133-40-6 EINECS: 218-363-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...