4-Brom-1-(4-methylphenyl)-1H-pyrazol 98%

APOSOR11501-1GEA 0 EUR
APOSOR11501-1G APOSOR11501-5G
4-Brom-1-(4-methylphenyl)-1H-pyrazol 98%
4-Bromo-1-(4-methylphenyl)-1H-pyrazole
Formel: C₁₀H₉BrN₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09475854
CAS-Nummer: 957034-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR