3-Chlor-6-(4-methyl-1-imidazolyl)pyridazin 98%
|
Achtung
|
APOSOR11502-1GEA
118
EUR
APOSOR11502-1G
APOSOR11502-5G
3-Chlor-6-(4-methyl-1-imidazolyl)pyridazin 98%
3-Chloro-6-(4-methyl-1-imidazolyl)pyridazine
Formel:
C₈H₇ClN₄ Molekulargewicht: 194,6210 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD09800928 CAS-Nummer: 941294-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...