4-Chlor-2-methoxy-N-methylanilin 96%
|
Achtung
|
APOSOR11538-250MGEA
0
EUR
APOSOR11538-250MG
APOSOR11538-1G
4-Chlor-2-methoxy-N-methylanilin 96%
4-Chlor-2-methoxy-N-methylanilin
Formel:
C₈H₁₀ClNO Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00027506 CAS-Nummer: 35122-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...