4-Brom-N-(furan-2-ylmethyl)benzamid

Achtung

APOSOR11620-1GEA 0 EUR
APOSOR11620-1G APOSOR11620-5G
4-Brom-N-(furan-2-ylmethyl)benzamid
4-Brom-N-(furan-2-ylmethyl)benzamid
Formel: C₁₂H₁₀BrNO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00451751
CAS-Nummer: 312587-75-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR