2-Brom-N-(3-methoxypropyl)benzolsulfonamid

Lieferant: Apollo Scientific

APOSOR11642-1GEA 132 EUR
APOSOR11642-1G APOSOR11642-5G
2-Brom-N-(3-methoxypropyl)benzolsulfonamid
2-Brom-N-(3-methoxypropyl)benzolsulfonamid
Formel: C₁₀H₁₄BrNO₃S
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07710755
CAS-Nummer: 848906-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR