3-Chlor-6-(ethylamino)pyridazin

APOSOR11900-5GEA 0 EUR
APOSOR11900-5G APOSOR11900-1G
3-Chlor-6-(ethylamino)pyridazin
3-Chloro-6-(ethylamino)pyridazine
Formel: C₆H₈ClN₃
Molekulargewicht: 157,6008 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08363097
CAS-Nummer: 68588-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR