Methyl-5-brom-4-chlor-2-methoxybenzoat
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR11905-1GEA
46.6
EUR
APOSOR11905-1G
Methyl-5-brom-4-chlor-2-methoxybenzoat
Methyl-5-brom-4-chlor-2-methoxybenzoat
Formel:
C₉H₈BrClO₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09800923 CAS-Nummer: 951885-11-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...