3-Brom-N-cyclopentylbenzamid

APOSOR11924-1GEA 0 EUR
APOSOR11924-1G APOSOR11924-5G
3-Brom-N-cyclopentylbenzamid
3-Brom-N-cyclopentylbenzamid
Formel: C₁₂H₁₄BrNO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00751460
CAS-Nummer: 349405-34-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR