2-Amino-4-(3-pyridyl)thiazol
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR1300-1GEA
87.3
EUR
APOSOR1300-1G
2-Amino-4-(3-pyridyl)thiazol
2-Amino-4-(3-pyridyl)thiazole
Formel:
C₈H₇N₃S Molekulargewicht: 177,22 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00218013 CAS-Nummer: 30235-27-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...