Methyl-2-amino-4-methylthiazol-5-carboxylat

Achtung

APOSOR13047-5GEA 0 EUR
APOSOR13047-5G APOSOR13047-25G
Methyl-2-amino-4-methylthiazol-5-carboxylat
Methyl-2-amino-4-methylthiazol-5-carboxylat
Formel: C₆H₈N₂O₂S
Molekulargewicht: 172,2049 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD01993690
CAS-Nummer: 3829-80-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR