1-(3-Aminophenylsulfonyl)pyrrolidin
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR14250-5GEA
408
EUR
APOSOR14250-5G
APOSOR14250-1G
1-(3-Aminophenylsulfonyl)pyrrolidin
1-(3-Aminophenylsulfonyl)pyrrolidin
Formel:
C₁₀H₁₄N₂O₂S Schmelzpunkt: 155…156 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02615287 CAS-Nummer: 91619-38-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...