2-(Methylsulfonyl)-4'-nitroacetophenon
APOSOR14844-5GEA
0
EUR
APOSOR14844-5G
APOSOR14844-1G
APOSOR14844-10G
2-(Methylsulfonyl)-4'-nitroacetophenon
2-(Methylsulfonyl)-4'-nitroacetophenon
Formel:
C₉H₉NO₅S Schmelzpunkt: 160…167 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09865011 CAS-Nummer: 136819-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...