2-(4-Piperidinyl)benzoxazol

Achtung

APOSOR15240-5GEA 0 EUR
APOSOR15240-5G APOSOR15240-10G APOSOR15240-1G
2-(4-Piperidinyl)benzoxazol
2-(4-Piperidinyl)benzoxazol
Formel: C₁₂H₁₄N₂O
Schmelzpunkt: 114…116 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01475844
CAS-Nummer: 51784-03-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR