2-(Methylsulfonyl)-5-(tributylstannyl)pyrimidin

APOSOR15526-500MGEA 390 EUR
APOSOR15526-500MG
2-(Methylsulfonyl)-5-(tributylstannyl)pyrimidin
2-(Methylsulfonyl)-5-(tributylstannyl)pyrimidin
Formel: C₁₇H₃₂N₂O₂SSn
Molekulargewicht: 447,22 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07787379
CAS-Nummer: 122476-85-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR