(3R)-3-Pyrrolidinolhydrochlorid

APOSOR15681-25GEA 0 EUR
APOSOR15681-25G
(3R)-3-Pyrrolidinolhydrochlorid
(R)-3-Hydroxypyrrolidine hydrochloride
Formel: C₄H₁₀ClNO
Molekulargewicht: 123,58 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00191570
CAS-Nummer: 104706-47-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR