7-Nitroindole-3-carboxyaldehyde 95+%

Lieferant: Apollo Scientific
Achtung

APOSOR1670-1GEA 20.5 EUR
APOSOR1670-1G
7-Nitroindole-3-carboxyaldehyde 95+%
7-Nitroindole-3-carboxyaldehyde
Formel: C₉H₆N₂O₃
Molekulargewicht: 190,16 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD01074519
CAS-Nummer: 10553-14-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR