5-Pyrimidineacetic acid
APOSOR17835-250MGEA
0
EUR
APOSOR17835-250MG
APOSOR17835-1G
APOSOR17835-5G
5-Pyrimidineacetic acid
5-Pyrimidineacetic acid
Formel:
C6H6N2O2 Molekulargewicht: 138,1240 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09608122 CAS-Nummer: 5267-07-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...