3-Amino-6-brom-2-methoxypyridin
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR18103-250MGEA
19
EUR
APOSOR18103-250MG
3-Amino-6-brom-2-methoxypyridin
3-Amino-6-bromo-2-methoxypyridine
Formel:
C₆H₇BrN₂O Molekulargewicht: 203,0400 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06738312 CAS-Nummer: 89466-18-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...