5-Amino-3-methyl-1-phenyl-1H-pyrazol 98%
Lieferant: Apollo Scientific
APOSOR22602-100GEA
65.5
EUR
APOSOR22602-100G
5-Amino-3-methyl-1-phenyl-1H-pyrazol 98%
5-Amino-3-methyl-1-phenyl-1H-pyrazole
Formel:
C₁₀H₁₁N₃ Molekulargewicht: 173,21 g/mol Schmelzpunkt: 113…115 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00020727 CAS-Nummer: 1131-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...