1,2,3,4,6,7-Hexabromnaphthalin

Achtung

APOSOR22848-250MGEA 0 EUR
APOSOR22848-250MG
1,2,3,4,6,7-Hexabromnaphthalin
1,2,3,4,6,7-Hexabromonaphthalene
Formel: C₁₀H₂Br₆
Lagerungstemperatur: Raumtemperatur
CAS-Nummer: 75625-24-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR