3-(Hydroxymethyl)benzonitril
Lieferant: Apollo Scientific
APOSOR301001-1GEA
53.9
EUR
APOSOR301001-1G
3-(Hydroxymethyl)benzonitril
3-(Hydroxymethyl)benzonitril
Formel:
C₈H₇NO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01321087 CAS-Nummer: 874-97-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...