1-tert-Butyl-3-ethylpyrrolidine-1,3-dicarboxylate

Lieferant: Apollo Scientific

APOSOR301175-5GEA 313 EUR
APOSOR301175-5G
1-tert-Butyl-3-ethylpyrrolidine-1,3-dicarboxylate
1-tert-Butyl-3-ethylpyrrolidine-1,3-dicarboxylate
Formel: C₁₂H₂₁NO₄
Molekulargewicht: 243,30
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03791256
CAS-Nummer: 170844-49-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR