4-(Piperidin-1-carbonyl)phenylboronsäurepinakolester

APOSOR303400-1GEA 0 EUR
APOSOR303400-1G APOSOR303400-10G APOSOR303400-5G
4-(Piperidin-1-carbonyl)phenylboronsäurepinakolester
4-(Piperidin-1-carbonyl)phenylboronsäurepinakolester
Formel: C₁₈H₂₆BNO₃
Molekulargewicht: 315,21 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09027083
CAS-Nummer: 938043-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR