4'-Methoxybiphenyl-4-ylboronsäurepinakolester
APOSOR303484-1GEA
0
EUR
APOSOR303484-1G
APOSOR303484-5G
APOSOR303484-500MG
4'-Methoxybiphenyl-4-ylboronsäurepinakolester
4'-Methoxybiphenyl-4-ylboronsäurepinakolester
Formel:
C₁₉H₂₃BO₃ Molekulargewicht: 310,20 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD16294530 CAS-Nummer: 446311-34-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...