2-Methyl-2H-indazol-6-boronsäurepinakolester
|
Achtung
|
APOSOR30598-1GEA
0
EUR
APOSOR30598-1G
2-Methyl-2H-indazol-6-boronsäurepinakolester
2-Methyl-2H-indazol-6-boronsäurepinakolester
Formel:
C₁₄H₁₉BN₂O₂ Molekulargewicht: 258,1239 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD11109435 CAS-Nummer: 1204580-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...