6-Brom-5-methyl-1H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR30743-1GEA
81.5
EUR
APOSOR30743-1G
6-Brom-5-methyl-1H-indazol
6-Bromo-5-methyl-1H-indazole
Formel:
C₈H₇BrN₂ Molekulargewicht: 211,0600 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD09026997 CAS-Nummer: 1000343-69-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...