6-Bromchinolin-2(1H)-on 98%
|
Achtung
|
APOSOR307506-1GEA
0
EUR
APOSOR307506-1G
APOSOR307506-25G
APOSOR307506-5G
6-Bromchinolin-2(1H)-on 98%
6-Bromchinolin-2(1H)-on
Formel:
C₉H₆BrNO Molekulargewicht: 224,05 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD08703169 CAS-Nummer: 1810-66-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...