1-Methyl-1H-indazol-5-boronsäurepinakolester
APOSOR308051-1GEA
0
EUR
APOSOR308051-1G
APOSOR308051-5G
1-Methyl-1H-indazol-5-boronsäurepinakolester
1-Methyl-1H-indazol-5-boronsäurepinakolester
Formel:
C₁₄H₁₉BN₂O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD15071438 CAS-Nummer: 1235469-00-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...