Benzyl-(R)-(-)-glycidylether 98%
|
Achtung
|
APOSOR315270-100GEA
0
EUR
APOSOR315270-100G
APOSOR315270-25G
Benzyl-(R)-(-)-glycidylether 98%
Benzyl-(R)-(-)-glycidylether
Formel:
C₁₀H₁₂O₂ Molekulargewicht: 164,20 g/mol Dichte: 1,077 g/cm³ (25 °C) Flammpunkt: 110 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00068409 CAS-Nummer: 14618-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...