4-Amino-3-methoxybenzonitril 98%

Lieferant: Apollo Scientific

APOSOR321008-1GEA 74.2 EUR
APOSOR321008-1G
4-Amino-3-methoxybenzonitril 98%
4-Amino-3-methoxybenzonitril
Formel: C₈H₈N₂O
Molekulargewicht: 148,1600 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD13250177
CAS-Nummer: 177476-76-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR