N-[(Benzyloxy)carbonyl]serin
Lieferant: Apollo Scientific
APOSOR322046-5GEA
49.5
EUR
APOSOR322046-5G
N-[(Benzyloxy)carbonyl]serin
N-Cbz-L-serine
Formel:
C₁₁H₁₃NO₅ Molekulargewicht: 239,2 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00002662 CAS-Nummer: 1145-80-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...